Benzofuran-2-carboxaldehyde 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzo[b]furan-2-carboxaldehyde, Benzofuran-2-carbaldehyde

33014-0010 33014-0050
TS33014-0010EA 67 USD
TS33014-0010 TS33014-0050
Benzofuran-2-carboxaldehyde 96%
Benzofuran-2-carboxaldehyde
Formula: C₉H₆O₂
MW: 146.145 g/mol
Density: 1.206 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015463
CAS Number: 4265-16-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6300 to 1.6340 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR