6,7-Dihydro-4(5H)-benzofuranone 98%

Supplier: Thermo Scientific Chemicals

33008-0050 33008-0250
TS33008-0050EA 112 USD
TS33008-0050 TS33008-0250
6,7-Dihydro-4(5H)-benzofuranone 98%
6,7-Dihydro-4(5H)-benzofuranone
Formula: C₈H₈O₂
Boiling Pt: 115…118 °C (16 mmHg)
Melting Pt: 30…34 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00215511
CAS Number: 16806-93-2

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR