5-Bromo-2-furaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

32952-0010 32952-0100
TS32952-0010EA 28 USD
TS32952-0010 TS32952-0100
5-Bromo-2-furaldehyde 97%
5-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 201 °C (760 mmHg)
Melting Pt: 80…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159501
CAS Number: 1899-24-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 80 to 85 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR