Ethyl-2-methylacetoacetate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 2-methyl-3-oxobutanoate

32611-0250 32611-1000
TS32611-0250EA 49 USD
TS32611-0250 TS32611-1000
Ethyl-2-methylacetoacetate 95%
Ethyl-2-methylacetoacetate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.01 g/cm³
Flash Pt: 62 °C
MDL Number: MFCD00009164
CAS Number: 609-14-3
EINECS: 210-179-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4230 (20 °C, 589 nm)
Water ≤1 % (K.F.)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR