Diethyl-3-oxoglutarate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

32563-0500 32563-2500
TS32563-0500EA 58 USD
TS32563-0500 TS32563-2500
Diethyl-3-oxoglutarate 95%
Diethyl-3-oxoglutarate
Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009200
CAS Number: 105-50-0
EINECS: 203-302-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4420 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR