trans-2-Pentenal 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
32542-0050
32542-0250
TS32542-0050EA
42
USD
TS32542-0050
TS32542-0250
trans-2-Pentenal 97%
trans-2-Pentenal
Formula:
C₅H₈O Boiling Pt: 127 °C (1013 hPa) Density: 0.86 g/cm³ Flash Pt: 19 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009615 CAS Number: 1576-87-0 EINECS: 216-414-1 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4400 to 1.4470 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...