Methyl-β-D-galactopyranoside 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

31498-0010 31498-0050
TS31498-0010EA 75 USD
TS31498-0010 TS31498-0050
Methyl-β-D-galactopyranoside 98%
Methyl-beta-D-galactopyranoside
Formula: C₇H₁₄O₆
Melting Pt: 175…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064357
CAS Number: 1824-94-8

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥97.5 % (ELSD)
Specific optical rotation -15° to -19° (20°C, 589 nm) (c=1.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR