cis-1,2-Cyclopentanediol 98%

Supplier: Thermo Scientific Chemicals

31029-0010 31029-0050
TS31029-0010EA 51 USD
TS31029-0010 TS31029-0050
cis-1,2-Cyclopentanediol 98%
cis-1,2-Cyclopentanediol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 83…83.5 °C (5 mmHg)
Melting Pt: 32…33 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134192
CAS Number: 5057-98-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4790 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR