5-Bromo-1-indanone 97%

Supplier: Thermo Scientific Chemicals
Warning

30957-0010 30957-0050
TS30957-0010EA 38 USD
TS30957-0010 TS30957-0050
5-Bromo-1-indanone 97%
5-Bromo-1-indanone
Formula: C₉H₇BrO
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now

Specification Test Results

Appearance White to beige to brown Powder
Infrared spectrum Conforms
Melting point 125 to 130 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR