4'-tert-Butylacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30874-0010
30874-0050
TS30874-0010EA
32
USD
TS30874-0010
TS30874-0050
4'-tert-Butylacetophenone 98%
4'-tert-Butylacetophenone
Formula:
C₁₂H₁₆O Boiling Pt: 107…108 °C (5 mmHg) Melting Pt: 17…18 °C Density: 0.96 g/cm³ Flash Pt: 29 °C |
MDL Number:
MFCD00017256 CAS Number: 943-27-1 EINECS: 213-399-3 UN: 1224 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5190 to 1.5220 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...