1-Bromo-3,3-dimethylbutanone 93%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone

30641-0050 30641-0250
TS30641-0050EA 47 USD
TS30641-0050 TS30641-0250
1-Bromo-3,3-dimethylbutanone 93%
1-Bromo-3,3-dimethylbutanone
Formula: C₆H₁₁BrO
MW: 179.06 g/mol
Boiling Pt: 189…192 °C (760 mmHg)
Melting Pt: –10 °C
Density: 1.33 g/cm³
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000206
CAS Number: 5469-26-1
EINECS: 226-794-0

Order Now

Specification Test Results

Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4656 to 1.4676 (20 °C, 589 nm)
GC ≥91 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR