ω-Bromo-4-fluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

30629-0050 30629-0250
TS30629-0050EA 55 USD
TS30629-0050 TS30629-0250
ω-Bromo-4-fluoroacetophenone 98%
ω-Bromo-4-fluoroacetophenone
Formula: C₈H₆BrFO
Boiling Pt: 150 °C (12 mmHg)
Melting Pt: 46…49 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow to yellow-green to beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 45 to 50 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR