ω-Bromo-4-nitroacetophenone 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

30548-0100 30548-0500
TS30548-0100EA 53 USD
TS30548-0100 TS30548-0500
ω-Bromo-4-nitroacetophenone 95%
ω-Bromo-4-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 94…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 100 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR