4,4'-Diacetylbiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning

30544-0010 30544-0050
TS30544-0010EA 31 USD
TS30544-0010 TS30544-0050
4,4'-Diacetylbiphenyl 98%
4,4'-Diacetylbiphenyl
Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00017248
CAS Number: 787-69-9
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance Colorless to tan Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 197 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR