cis-1,2-Cyclohexanediol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: cis-Cyclohexane-1,2-diol

30097-0010 30097-0050
TS30097-0010EA 50 USD
TS30097-0010 TS30097-0050
cis-1,2-Cyclohexanediol 99%
cis-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00064944
CAS Number: 1792-81-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or flakes
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR