cis,cis-Muconic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2Z,4Z)-Hexa-2,4-dienedioic acid

29776-0025 29776-0100
TS29776-0025EA 103 USD
TS29776-0025 TS29776-0100
cis,cis-Muconic acid 98%
cis,cis-Muconic acid
Formula: C₆H₆O₄
MW: 142.11 g/mol
Melting Pt: 194…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00085266
CAS Number: 1119-72-8

Order Now

Specification Test Results

Appearance Yellow to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR