β-Ionone (synthetic) 96%
Supplier: Thermo Scientific Chemicals
|
|
Synonyms:
(E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone
29713-0050
29713-1000
29713-5000
TS29713-0050EA
31
USD
TS29713-0050
TS29713-1000
TS29713-5000
β-Ionone (synthetic) 96%
(E)-β-Ionone
Formula:
C₁₃H₂₀O MW: 192.30 g/mol Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001549 CAS Number: 79-77-6 EINECS: 201-224-3 UN: 3082 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5190 to 1.5215 (20 °C, 589 nm) |
GC | ≥95 % |
Specific gravity | (25°C) 0.941 to 0.947 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...