3-Formylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

29656-0010 29656-0050
TS29656-0010EA 54 USD
TS29656-0010 TS29656-0050
3-Formylbenzoic acid 98%
3-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00039575
CAS Number: 619-21-6
EINECS: 210-585-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 172 to 176 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR