3-Buten-1-ol 98+%
Supplier: Thermo Scientific Chemicals
29575-0250
29575-1000
29575-5000
TS29575-0250EA
143
USD
TS29575-0250
TS29575-1000
TS29575-5000
3-Buten-1-ol 98+%
3-Buten-1-ol
Formula:
C₄H₈O |
MDL Number:
MFCD00002959 CAS Number: 627-27-0 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4200 to 1.4220 (20 °C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...