4-Quinolinecarbaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde

29482-0010 29482-0050 29482-0250
TS29482-0010EA 33 USD
TS29482-0010 TS29482-0050 TS29482-0250
4-Quinolinecarbaldehyde 95%
4-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 45…52 °C
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0

Order Now

Specification Test Results

Appearance Off-white to yellow to light brown Crystalline powder and/or chunks or solid
Infrared spectrum Conforms
Melting point 45 to 52 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR