3,4-Hexanedione 96%

Supplier: Thermo Scientific Chemicals
Warning

29327-0250 29327-2500
TS29327-0250EA 42 USD
TS29327-0250 TS29327-2500
3,4-Hexanedione 96%
3,4-Hexanedione
Formula: C₆H₁₀O₂
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.941 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00010237
CAS Number: 4437-51-8
EINECS: 224-651-7
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4150 (20 °C, 589 nm)
GC ≥95 %
Specific gravity (25°C) 0.93 to 0.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR