Isobornyl methacrylate 85-90% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate, (1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate(relative), Methacrylic acid Isobornyl Ester

29209-0010 29209-2500
TS29209-0010EA 212 USD
TS29209-0010 TS29209-0500 TS29209-2500
Isobornyl methacrylate 85-90% stabilized
Isobornyl methacrylate
Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: –60 °C
Density: 0.98 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00081070
CAS Number: 7534-94-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20 °C, 589 nm)
GC ≥85 %
Stabilizer 100 to 200 ppm MEHQ

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR