(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol

27955-0025 27955-0100
TS27955-0025EA 49 USD
TS27955-0025 TS27955-0100
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 98%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Melting Pt: 52…60 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077851
CAS Number: 18680-27-8

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals or mass
Infrared spectrum Conforms
Melting point 52 to 60 °C
GC ≥97.5 %
Specific optical rotation +7° to +10° (20 °C, 589 nm) (c=6.5, toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR