Octanophenone 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: n-Octanophenone

27936-0050 27936-0250 27936-1000
TS27936-0050EA 50 USD
TS27936-0050 TS27936-0250 TS27936-1000
Octanophenone 99+%
Octanophenone
Formula: C₆H₅C(O)(CH₂)₆CH₃
MW: 204.31 g/mol
Boiling Pt: 285…290 °C (760 mmHg)
Melting Pt: 21.5…23.5 °C
Density: 0.93 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003554
CAS Number: 1674-37-9
EINECS: 216-817-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5024 to 1.5054 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR