4'-Iodoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

27889-0050 27889-0250
TS27889-0050EA 35 USD
TS27889-0050 TS27889-0250
4'-Iodoacetophenone 98%
4'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
Melting Pt: 82…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 89 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR