3'-Chloropropiophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one

27658-0250 27658-2500
TS27658-0250EA 29 USD
TS27658-0250 TS27658-2500
3'-Chloropropiophenone 98%
3'-Chloropropiophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 124 °C (14 mmHg)
Melting Pt: 43…47 °C
Flash Pt: >110 °C
MDL Number: MFCD00009925
CAS Number: 34841-35-5
EINECS: 252-242-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR