Metaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Metacetaldehyde, Aldehyde (meta), Acetaldehyde Tetramer

27186-0010 27186-0050 27186-2500
TS27186-0010EA 348 USD
TS27186-0010 TS27186-0050 TS27186-2500
Metaldehyde 99%
Metaldehyde
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Melting Pt: 246 °C
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134115
CAS Number: 108-62-3
UN: 1332
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR