Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
27181-0500
27181-1000
27181-5000
TS27181-0500EA
32
USD
TS27181-0500
TS27181-1000
TS27181-5000
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 99+%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
Formula:
C₁₀H₁₂O₆ MW: 228.2 g/mol Melting Pt: 154…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001607 CAS Number: 6289-46-9 EINECS: 228-528-9 |
Specification Test Results
Appearance | White to yellow-green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | ≥153 °C |
HPLC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...