6-Bromo-1-hexanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane

27009-0025 27009-0100 27009-0500
TS27009-0025EA 59 USD
TS27009-0025 TS27009-0100 TS27009-0500
6-Bromo-1-hexanol 95%
6-Bromo-1-hexanol
Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 104…106 °C (5 mmHg)
Density: 1.32 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4810 to 1.4840 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR