3-Phenyl-2-propyn-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Phenylprop-2-yn-1-ol
27003-0050
27003-0250
TS27003-0050EA
67
USD
TS27003-0050
TS27003-0250
3-Phenyl-2-propyn-1-ol 98%
3-Phenyl-2-propyn-1-ol
Formula:
C₉H₈O MW: 132.16 g/mol Boiling Pt: 129…130 °C (10 mmHg) Density: 1.078 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040914 CAS Number: 1504-58-1 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5830 to 1.5870 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...