Bis(2-ethylhexyl)sebacate 97%

Supplier: Thermo Scientific Chemicals

26992-0010 26992-0025 26992-0100
TS26992-0010EA 83 USD
TS26992-0010 TS26992-0025 TS26992-0100
Bis(2-ethylhexyl)sebacate 97%
Bis(2-ethylhexyl)sebacate
Formula: C₂₆H₅₀O₄
Boiling Pt: 212 °C (1 mmHg)
Melting Pt: –55 °C
Density: 0.91 g/cm³
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00009497
CAS Number: 122-62-3
EINECS: 204-558-8
Merck Index: 13,01251

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4550 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR