(S)-(+)-2-Hydroxyisovaleric acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
26990-0010
26990-0050
TS26990-0010EA
65
USD
TS26990-0010
TS26990-0050
(S)-(+)-2-Hydroxyisovaleric acid 99+%
(S)-(+)-2-Hydroxyisovaleric acid
Formula:
C₅H₁₀O₃ Melting Pt: 65…68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066443 CAS Number: 17407-55-5 |
Specification Test Results
Appearance | White Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 65 to 70 °C |
GC | ≥99.0 % |
Specific optical rotation | +15° to +19° (24 °C, 589 nm) (c=2, CHCl3) |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...