1-Amino-3,3-diethoxypropane 97%

Supplier: Thermo Scientific Chemicals
Danger

26985-0250 26985-1000
TS26985-0250EA 171 USD
TS26985-0250 TS26985-1000
1-Amino-3,3-diethoxypropane 97%
1-Amino-3,3-diethoxypropane
Formula: C₇H₁₇NO₂
Boiling Pt: 72 °C (16hPa)
Density: 0.91 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00142581
CAS Number: 41365-75-7
UN: 2735
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4233 to 1.4253 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR