Ethyl-3,3-diethoxypropionate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3,3-diethoxypropanoate, Ethyl 3,3-diethoxypropionate

26964-0050 26964-0250 26964-1000
TS26964-0050EA 35 USD
TS26964-0050 TS26964-0250 TS26964-1000
Ethyl-3,3-diethoxypropionate 98%
Ethyl-3,3-diethoxypropionate
Formula: C₉H₁₈O₄
Boiling Pt: 95 °C (17 mmHg)
Density: 0.97 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009865
CAS Number: 10601-80-6
EINECS: 234-223-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR