Potassium methyl malonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

26955-0250 26955-1000 26955-5000
TS26955-0250EA 69 USD
TS26955-0250 TS26955-1000 TS26955-5000
Potassium methyl malonate 98%
Potassium methyl malonate
Formula: C₄H₅KO₄
MW: 156.18 g/mol
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR