cis-2-Aminomethyl-1-cyclohexanol hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

26588-0010 26588-2500
TS26588-0010EA 337 USD
TS26588-0010 TS26588-2500
cis-2-Aminomethyl-1-cyclohexanol hydrochloride 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride
Formula: C₇H₁₅NO·HCl
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00143984
CAS Number: 24947-68-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR