3-Methoxy-3-methyl-1-butanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-3-methoxybutanol, Solvant 39, MMB, 3-Methoxy-3-methylbutan-1-ol

25975-0010 25975-2500
TS25975-0010EA 135 USD
TS25975-0010 TS25975-2500
3-Methoxy-3-methyl-1-butanol 99%
3-Methoxy-3-methyl-1-butanol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 174 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.926 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR