1-Amino-3-methoxypropane 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methoxypropylamine

25752-0010 25752-0050
TS25752-0010EA 50 USD
TS25752-0010 TS25752-0050
1-Amino-3-methoxypropane 99+%
1-Amino-3-methoxypropane
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.87 g/cm³
Flash Pt: 48 °C
MDL Number: MFCD00014831
CAS Number: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4185 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR