3-Methoxypropiononitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

25744-0025 25744-1000 25744-5000
TS25744-0025EA 339 USD
TS25744-0025 TS25744-1000 TS25744-5000
3-Methoxypropiononitrile 98%
3-Methoxypropiononitrile
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 164…165 °C (760 mmHg)
Melting Pt: –62.9 °C
Density: 0.938 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR