3,3-Dimethylbutyraldehyde

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dimethylbutanal

25726-0010 25726-0050 25726-0250
TS25726-0010EA 66 USD
TS25726-0010 TS25726-0050 TS25726-0250
3,3-Dimethylbutyraldehyde
3,3-Dimethylbutyraldehyde
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 104…106 °C (760 mmHg)
Melting Pt: –24 °C
Density: 0.798 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00042807
CAS Number: 2987-16-8
UN: 1989
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR