2-Acetyl-1H-pyrrole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

25249-0050 25249-0250
TS25249-0050EA 40 USD
TS25249-0050 TS25249-0250
2-Acetyl-1H-pyrrole 99%
2-Acetyl-1H-pyrrole
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 91 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR