meso-Hydrobenzoin 95%

Supplier: Thermo Scientific Chemicals

Synonyms: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol

24768-0050 24768-0250
TS24768-0050EA 82 USD
TS24768-0050 TS24768-0250
meso-Hydrobenzoin 95%
meso-Hydrobenzoin
Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Melting Pt: 136…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00064253
CAS Number: 579-43-1

Order Now

Specification Test Results

Appearance White Crystalline powder and or lumps
Infrared spectrum Conforms
Melting point 134 to 140 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR