2',4'-Difluoroacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,4-Difluorophenyl)ethan-1-one

24677-0100 24677-0500
TS24677-0100EA 105 USD
TS24677-0100 TS24677-0500
2',4'-Difluoroacetophenone 99%
2',4'-Difluoroacetophenone
Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 80…81 °C (25 mmHg)
Density: 1.23 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00151261
CAS Number: 364-83-0
EINECS: 206-667-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4890 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR