Methyl propionylacetate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

24316-0250 24316-1000 24316-5000
TS24316-0250EA 49 USD
TS24316-0250 TS24316-1000 TS24316-5000
Methyl propionylacetate 99+%
Methyl propionylacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 95…97 °C (48 mmHg)
Melting Pt: –35 °C
Density: 1.03 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00011705
CAS Number: 30414-53-0
EINECS: 250-184-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20 °C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR