1,2-Difluoro-4-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Difluoronitrobenzene

24193-0250 24193-1000
TS24193-0250EA 82 USD
TS24193-0250 TS24193-1000
1,2-Difluoro-4-nitrobenzene 99%
1,2-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80…81 °C (14 mmHg)
Density: 1.441 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR