Methyl-4-chloroacetoacetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 4-chloro-3-oxobutanoate

24162-0250 24162-1000
TS24162-0250EA 59 USD
TS24162-0250 TS24162-1000
Methyl-4-chloroacetoacetate 97%
Methyl-4-chloroacetoacetate
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 212…213 °C (760 mmHg)
Melting Pt: 14…16 °C
Density: 1.28 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000938
CAS Number: 32807-28-6
EINECS: 251-230-5
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear light yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR