1,4-Dioxaspiro[4.5]decan-8-ol 98%

Supplier: Thermo Scientific Chemicals

24101-0010 24101-0050
TS24101-0010EA 90 USD
TS24101-0010 TS24101-0050
1,4-Dioxaspiro[4.5]decan-8-ol 98%
1,4-Dioxaspiro[4.5]decan-8-ol
Formula: C₈H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00067003
CAS Number: 22428-87-1

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4870 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR