(4-Bromophenyl)(cyclopropyl)ketone 95%

Supplier: Thermo Scientific Chemicals
Warning

23542-0500 23542-2500
TS23542-0500EA 193 USD
TS23542-0500 TS23542-2500
(4-Bromophenyl)(cyclopropyl)ketone 95%
(4-Bromophenyl)(cyclopropyl)ketone
Formula: C₁₀H₉BrO
Density: 1.45 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00019227
CAS Number: 6952-89-2

Order Now

Specification Test Results

Appearance Yellow to beige to orange Crystalline powder or low melting crystalline mass
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR