4-Benzoylpiperidine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenyl(piperidin-4-yl)methanone hydrochloride

23286-0010 23286-0100
TS23286-0010EA 42 USD
TS23286-0010 TS23286-0100
4-Benzoylpiperidine hydrochloride 98%
4-Benzoylpiperidine hydrochloride
Formula: C₁₂H₁₆ClNO
Melting Pt: 226…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00066982
CAS Number: 25519-80-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR