4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals

Synonyms: GLCNAC1-B-PNP, 4-Nitrophenyl N-Acetyl-β-D-glucosaminide, 4-Nitrophenyl 2-acetamido-2-deoxy-beta-D-glucopyranoside

22941-0010 22941-1000
TS22941-0010EA 243 USD
TS22941-0010 TS22941-1000
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside 99%
4-Nitrophenyl-2-acetamido-2-deoxy-beta-D-glucopyranoside
Formula: C₁₄H₁₈N₂O₈
MW: 342.30 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00063696
CAS Number: 3459-18-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (0.5 % in water) Clear colorless to light yellow
HPLC ≥98.5 %
Specific optical rotation -13° to -17° (20°C, 589 nm) (c=0.5, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR